EMAIL THIS PAGE TO A FRIEND
European journal of biochemistry
An aspartic endopeptidase is involved in the breakdown of propeptides of storage proteins in protein-storage vacuoles of plants.
PMID 9210475
Abstract
To understand the mechanism of the maturation of various proteins in protein-storage vacuoles, we purified a 48-kDa aspartic endopeptidase composed of 32-kDa and 16-kDa subunits from castor bean. Immunocytochemical and cell fractionation analyses of the endosperm of maturing castor bean seed showed that the aspartic endopeptidase was localized in the matrix of the protein-storage vacuoles, where a variety of seed storage proteins were also present. The amount of the aspartic endopeptidase increased at the mid-maturation stage of the seeds before accumulation of the storage proteins. To determine how the aspartic endopeptidase is responsible for maturation of seed proteins in concert with the vacuolar processing enzyme, we prepared 35S-labeled proproteins of seed proteins from the endoplasmic reticulum fraction of pulse-labeled maturing endosperm and used the authentic proproteins as substrates for in vitro processing experiments. The purified aspartic endopeptidase was unable to convert any of three endosperm proproteins, pro2S albumin, proglobulin, and proricin, into their mature sizes, while the purified vacuolar processing enzyme could convert all three proproteins. We further examined the activity of aspartic endopeptidase on the cleavage of an internal propeptide of Arabidopsis pro2S albumin, which is known to be removed post-translationally. The aspartic endopeptidase cleaved the propeptide at three sites under acidic conditions. These results suggest that aspartic endopeptidase cannot directly convert pro2S albumin into the mature form, but it may play a role in trimming the C-terminal propeptides from the subunits that are produced by the action of the vacuolar processing enzyme.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P2032 Pepstatin A−Agarose, saline suspension