EMAIL THIS PAGE TO A FRIEND
Methods in enzymology
Determining NAD synthesis in erythrocytes.
PMID 9211316
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
N7764
Nicotinic acid mononucleotide
C11H14NO9P