EMAIL THIS PAGE TO A FRIEND
Acta physiologica Hungarica
Effect of hyperglycemia on the intestinal elimination of p-nitrophenol in the rat.
PMID 9219610
Abstract
Effect of hyperglycemia on the intestinal elimination of p-nitrophenol has been investigated in rats. Hyperglycemia was produced by a continuous i.v. infusion of glucose, p-nitrophenol was used as a model compound for the investigation of intestinal metabolism and excretion of drugs. Intestinal conjugation of p-nitrophenol with glucuronic acid did not change significantly in hyperglycemic rats, however, formation of sulfoconjugate of p-nitrophenol was enhanced by hyperglycemia. Sum of metabolites (p-nitrophenol glucuronide and sulfate) appeared in the intestinal lumen in hyperglycemic rats was similar to the total luminal appearance of these metabolites of control rats.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
N3877
Potassium 4-nitrophenyl sulfate, sulfatase substrate
C6H4KNO6S