EMAIL THIS PAGE TO A FRIEND
Journal of pharmaceutical and biomedical analysis
Selected applications of capillaries with dynamic or permanent anodal electroosmotic flow in chiral separations by capillary electrophoresis.
PMID 9226592
Abstract
The techniques for a dynamic and permanent reversal of the electroosmotic flow (EOF) were used for the reversal of the enantiomer migration order (EMO) of neutral and cationic analytes in chiral capillary electrophoresis (CE). Native beta-Cd and an anionic CD derivative, CM-beta-CD were used in both, bare silica- and positively coated capillaries. Advantages and disadvantages of a dynamic and permanent modification of the capillary inner surface are briefly discussed.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
180823
5-Methyl-5-phenylhydantoin, 99%
C10H10N2O2