EMAIL THIS PAGE TO A FRIEND
Cell
Hemoglobin synthesis in cell hybrids formed between teratocarcinoma and Friend erythroleukemia cells.
PMID 922887
Abstract
Viable hybrid cells have been isolated following fusion of Friend erythroleukemia cells and undifferentiated teratocarcinoma cells. The hybrids formed between near-diploid parental cells resembled Friend cells in the ability to grow in suspension and to synthesize hemoglobin in the presence of the chemical inducers dimethyl sulfoxide (DMSO) and ouabain. Erythropoietin (EPO) was effective in inducing hemoglobin synthesis in some of the hybrid cell lines. The hemoglobins synthesized by the hybrids were of the adult forms, but were quantitatively different from those hemoglobins synthesized by the parental Friend cells, suggesting that the fusion event modulated the expression of the hemoglobin chain genes.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
details
745C2, 87030901