EMAIL THIS PAGE TO A FRIEND
Journal of clinical microbiology
Simple color tests based on an alanyl peptidase reaction which differentiate Listeria monocytogenes from other Listeria species.
PMID 9230403
Abstract
The hydrolysis of DL-alanine-beta-naphthylamide and D-alanine-p-nitroanilide for identification of Listeria spp. has been studied with 227 cultures. All species of Listeria, except L. monocytogenes, hydrolyzed these substrates. The reactions were detected by simple chromogenic reactions and could substitute for the CAMP test.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A2503
DL-Alanine β-naphthylamide hydrochloride
C13H14N2O · HCl
A2628
L-Alanine β-naphthylamide, protease substrate
C13H14N2O