EMAIL THIS PAGE TO A FRIEND
Contact dermatitis
Allergic contact dermatitis from 2,7-dihydroxynaphthalene in hair dye.
PMID 9237018
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D116408
2,7-Dihydroxynaphthalene, 97%
C10H8O2