EMAIL THIS PAGE TO A FRIEND
Inflammation
Bradykinin antagonizes the effects of alpha-thrombin.
PMID 9246571
Abstract
alpha-Thrombin (AT) and bradykinin (BK) are endogenous mediators that are released during an inflammatory response, and could have a synergistic effect on endothelial permeability. Human umbilical vein endothelial cells (HUVEC) were grown on Transwell membranes and then tested for alterations in permeability to fluorescein isothiocyanate-labeled human serum albumin. Addition of 1 microM AT produced a significant increase in the permeability coefficient at 30 minutes from control levels of 1.59 x 10(-6) cm/sec to 4.92 x 10(-6) cm/sec. BK (1 microM) produced a similar increase to 4.46 x 10(-6) cm/sec. For both compounds, permeability remained elevated for 90 minutes. Pre-treatment of the HUVEC with the bradykinin receptor antagonist, Na-adamantaneacetyl-bradykinin (NA-BK) (1 microM), prior to addition of AT, reduced the AT permeability coefficient to 2.69 x 10(-6) cm/sec. Addition of NA-BK (1 microM) for 5 minutes, then BK (1 microM) for 5 minutes, inhibited the effect of BK and of AT (1 microM on permeability, decreasing the permeability coefficient of the endothelial monolayer to control levels (1.62 x 10(-6) cm/sec). AT (1 microM) increased HUVEC intracellular calcium mobilization, as monitored by FURA-2, to 245 nM from control (70 nM), however, pre-treatment with either BK or the bradykinin receptor antagonist decreased the AT induced intracellular calcium mobilization compared to AT alone. Pre-treatment of the HUVEC with bradykinin (1 microM) for 2 minutes also inhibited the effects of alpha-thrombin (1 microM) on f-actin distribution examined by BODIPY-phallodin staining and increased the clotting times for an alpha-thrombin dependent fibrinogen to fibrin clotting assay. However, incubation of bradykinin (1 microM) with alpha-thrombin (1 microM) for either 10 minutes or 100 minutes produced no detectable hydrolysis products. These data strongly suggest that the inflammatory mediators alpha-thrombin and bradykinin when released together, rather than being synergistic, are antagonistic.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
B1901
Bradykinin Fragment 2-9, ≥97% (HPLC)
C44H61N11O10
SCP0008 [DArg0,Hyp3,Thi5,8,DPhe7]-Adamantaneacetyl Bradykinin
C68H99N19O14S2