EMAIL THIS PAGE TO A FRIEND
Journal of Chromatography. B, Biomedical Sciences and Applications
Determination of 5-hydroxy-N-methylpyrrolidone and 2-hydroxy-N-methylsuccinimide in human urine.
PMID 9252049