EMAIL THIS PAGE TO A FRIEND
Journal of investigational allergology & clinical immunology
Recurrent fixed drug eruption caused by citiolone.
PMID 9252880
Abstract
Citiolone (N-acetylhomocysteinethiolactone) is a thiolic-derived medication frequently used in Spain and in other countries as a mucolytic agent for the treatment of certain hepatic disorders. Mucolytic drugs have rarely been implicated in the fixed drug eruption etiology. We report on a patient who presented several episodes of fixed exanthema related to citiolone intake. The patch test with citiolone (10% in dimethyl sulfoxide) was negative. The diagnosis was confirmed by a positive controlled oral challenge test. Other mucolytic thiolic-derivatives (N-acetylcysteine) were tolerated by the patient, thus crossreactivity between these drugs seems to be unlikely.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A16602
DL-N-Acetylhomocysteine thiolactone, 98%
C6H9NO2S