EMAIL THIS PAGE TO A FRIEND
Journal of chromatography. A
Analysis of benzalkonium chloride in the effluent from European hospitals by solid-phase extraction and high-performance liquid chromatography with post-column ion-pairing and fluorescence detection.
PMID 9253191
Abstract
A highly reproducible and specific method for the analysis of the quaternary ammonium compound, benzalkonium chloride, in effluents from European hospitals is presented. Benzalkonium chloride was extracted with end-capped RP-18 solid-phase cartridges and was selectively eluted. The resulting solution was analyzed by high-performance liquid chromatography (HPLC). After elution from the analytical column of the HPLC system, 9,10-dimethoxyanthracene-2-sulfonate was added continuously as a fluorescence marker, forming a hydrophobic ion-pair with benzalkonium chloride. The ion-pair was analyzed by fluorescence detection. The method was applied to highly complex effluent samples from different sized European hospitals. The measured concentrations were between 0.05 and 6.03 mg/l. The amounts emitted per bed and year were 4.5-362 g and did not correlate with the size of the hospital. The total amounts were 2.6-909 kg/year.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
38605
9,10-Dimethoxy-2-anthracenesulfonic acid sodium salt, for fluorescence, ≥96.0% (HPLC)
C16H13NaO5S