EMAIL THIS PAGE TO A FRIEND
Bulletin of environmental contamination and toxicology
Qualitative and quantitative analysis of diazinon in fabric exposed to various simulated sunlight and humidity conditions.
PMID 9256391