EMAIL THIS PAGE TO A FRIEND
Human mutation
Novel mutations of the tyrosinase (TYR) gene in type I oculocutaneous albinism (OCA1).
PMID 9259202
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T3824
Tyrosinase from mushroom, lyophilized powder, ≥1000 unit/mg solid