EMAIL THIS PAGE TO A FRIEND
Nutrition (Burbank, Los Angeles County, Calif.)
Effect of epidermal growth factor on glutamine metabolic enzymes in small intestine and skeletal muscle of parenterally fed rats.
PMID 9263258
Abstract
Exogenous epidermal growth factor (EGF) markedly increases the in vivo uptake of glutamine by small intestine during total parenteral nutrition (TPN). Since glutamine is the major oxidative fuel for the small intestine and is synthesized mainly in skeletal muscle, we investigated whether EGF would induce changes in the activity of the enzymes that mediate glutamine degradation (glutaminase) and synthesis (glutamine synthetase) in the two tissues. Male Sprague Dawley rats were randomized into three groups: group I (chow) were fed rat chow and water and libitum, group II (TPN) received a standard formula of TPN, and group III (TPN-EGF) received the same TPN as group II and injections of EGF (0.1 microgram/ gm body weight (bw)) subcutaneously twice daily. TPN was given for 2 wk; when EGF was administered along with TPN, the glutaminase activity of intestinal mucosa and the glutamine synthetase activity of skeletal muscle were increased, respectively, by 25% and 24% (P < 0.05, versus TPN group). These data suggest a mechanism whereby EGF enhances the intestinal utilization of glutamine by changing the activities of glutamine metabolic enzymes in the small intestine and skeletal muscle during TPN.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
G2253
L-Glutamic acid γ-monohydroxamate
C5H10N2O4