EMAIL THIS PAGE TO A FRIEND
Critical reviews in toxicology
Methoxychlor as a model for environmental estrogens.
PMID 9263644
Abstract
Estrogens can have a variety of physiological effects, especially on the reproductive system. Chemicals with estrogenic activity that are present in the environment may thus be considered potentially hazardous to development and/or reproduction. Methoxychlor is one such chemical, a chlorinated hydrocarbon pesticide with proestrogenic activity. Metabolism of the chemical either in vivo or using liver microsomes produces 2,2-bis(p-hydroxyphenyl)- 1,1,1-trichloroethane (HPTE), the active estrogenic form, and the delineation of this mechanism is reviewed herein. When administered in vivo, methoxychlor has adverse effects on fertility, early pregnancy, and in utero development in females as well as adverse effects on adult males such as altered social behavior following prenatal exposure to methoxychlor. Effects of methoxychlor on the female have been studied extensively, whereas reports on the chemical's effects on males are less common. From the studies reviewed here, the reproductive toxicity of methoxychlor is evident, but the significance of this toxicity with respect to human health remains to be determined.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
36161
Methoxychlor, PESTANAL®, analytical standard
C16H15Cl3O2
49054
Methoxychlor, analytical standard
C16H15Cl3O2