EMAIL THIS PAGE TO A FRIEND
Archives of andrology
Superoxide anion radical mediated dissection of rat sperm: an in vitro study.
PMID 9272225
Abstract
Sperm is highly sensitive to superoxide radical surge in the granuloma interior. To investigate possibility of a superoxide radical bomb, the role of an oxyradical on the spermatozoon was studied in vitro using potassium superoxide, a potent superoxide radical producer. Results confirm that the superoxide radical has a unique site-specific scissoring power.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
278904
Potassium dioxide, powder
KO2
310492
Potassium dioxide, chunks, 5-10 mm
KO2