EMAIL THIS PAGE TO A FRIEND
Life sciences
Growth suppression and induction of heat-shock protein-70 by 9-cis beta-carotene in cervical dysplasia-derived cells.
PMID 9275014
Abstract
The aim of the present study was to determine to what extent 9-cis beta-carotene, one of the most abundant naturally-occurring cis-isomers of beta-carotene, can inhibit the growth of cervical dysplasia-derived cells in comparison with all-trans beta-carotene. We found that 9-cis beta-carotene was dose-dependently more effective than all-trans beta-carotene. Both carotenes induced the intracellular accumulation of heat-shock protein-70 (HSP70), and the treated cells showed morphological changes indicative of apoptosis. The results of the present study strongly suggest that the induction of HSP70 by beta-carotene might be involved in beta-carotene-mediated suppression of the cell growth through apoptosis.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
52824
(9Z)-β-Carotene, ≥90.0% (HPLC)
C40H56