EMAIL THIS PAGE TO A FRIEND
Photochemistry and photobiology
Characterization of DNA damage inflicted by free radicals from a mutagenic sunscreen ingredient and its location using an in vitro genetic reversion assay.
PMID 9277149
Abstract
We describe an in vitro approach to assessing the potential genotoxicity of illuminated sunscreens. The photomutagenic sunscreen Padimate-O attacks DNA on illumination with simulated sunlight, producing strand breaks and lesions that are labile to N,N'-dimethylethylenediamine but few, if any, cyclobutane dimers or other direct photoproducts. The damage can be completely suppressed by the free radical quenchers Tris, ethanol, mannitol and dimethylsulfoxide, which is commonly used as a solvent in conventional photomutagenicity assays. Using a genetic reversion assay that depends on regenerating beta-galactosidase activity in photodamaged plasmids we find that GC base pairs are particularly susceptible to attack by Padimate-O.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
437395
2-Ethylhexyl 4-(dimethylamino)benzoate, 98%
C17H27NO2
74309
Padimate O, analytical standard
C17H27NO2