EMAIL THIS PAGE TO A FRIEND
Biulleten' eksperimental'noi biologii i meditsiny
[Evaluation of the level of lipid peroxidation during acute poisoning with organophosphorus pesticide in cats and the effect of antioxidants on their survival].
PMID 9280458
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
46424
Formothion solution, ~80% in xylene, PESTANAL®, analytical standard
C6H12NO4PS2