EMAIL THIS PAGE TO A FRIEND
Biological signals
Hypusine is essential for eukaryotic cell proliferation.
PMID 9285094
Abstract
Hypusine [N epsilon-(4-amino-2-hydroxybutyl)lysine] occurs in all eukaryotes at one residue in a highly conserved protein, the putative eukaryotic translation initiation factor 5A (eIF-5A, old terminology eIF-4D). This unusual amino acid is produced in a unique posttranslational modification reaction that involves the conjugation of the 4-aminobutyl moiety of the polyamine spermidine to the epsilon-amino group of a specific lysine residue of the eIF-5A precursor protein to form the deoxyhypusine [N epsilon-(4-aminobutyl)lysine] residue and its subsequent hydroxylation. The strict specificity of hypusine synthesis, its derivation from spermidine and its requirement for the activity of eIF-5A and for eukaryotic cell proliferation have raised keen interest in the physiological function of the hypusine-containing protein, eIF-5A.