EMAIL THIS PAGE TO A FRIEND
European journal of pharmacology
Competitive and non-competitive inhibition of L-3, 4-dihydroxyphenylalanine uptake in Opossum kidney cells.
PMID 9286625
Abstract
The present study aimed to determine the kinetics of L-3,4-dihydroxyphenylalanine (L-DOPA) uptake in Opossum kidney (OK) cells and to define the type of inhibition produced by L-5-hydroxytryptophan (L-5-HTP), cyanine 863 and 3,3'-diethyloxacarbocyanine (3,3'-DOC). Non-linear analysis of the saturation curves revealed for L-DOPA a Km (in microM) of 129 (114, 145) and a Vmax (in nmol/mg protein per 6 min) of 30.0 +/- 0.4 IC50 values for L-5-HTP (1454 microM) obtained in the presence of a nearly saturating (250 microM) concentration of L-DOPA were almost 4-fold those obtained when non-saturating (25 microM) concentrations of L-DOPA were used (330). IC50 values for cyanine 863 and 3,3'-DOC (638 and 353 microM) obtained in the presence of a nearly saturating (250 microM) concentration of L-DOPA were similar to those obtained when non-saturating (25 microM) concentrations of L-DOPA were used (654 and 339 microM). Vmax values (in nmol/mg protein per 6 min) for L-DOPA uptake were identical in the absence (36.4 +/- 0.7) and the presence of L-5-HTP (39.2 +/- 1.3), but Km values (microM) were significantly greater (P < 0.05) when L-DOPA uptake was studied in the presence of L-5-HTP (121 (100, 142) versus 318 (237, 399)). In contrast, the effect of cyanine 863 and 3,3'-DOC was to cause a significant reduction in Vmax values without significant changes in Km values. It is concluded that L-5-HTP exerts a competitive type of inhibition of L-DOPA uptake in cultured OK cells, whereas both cyanine 863, an organic cation transport inhibitor and 3,3'-DOC behave as non-competitive inhibitors.