EMAIL THIS PAGE TO A FRIEND
The Journal of biological chemistry
Nitroxides increase the detectable amount of nitric oxide released from endothelial cells.
PMID 9287307
Abstract
Nitroxides are known to exert superoxide dismutase-mimetic properties and to decrease O-2- and H2O2-mediated cytotoxicity. However, the effect of nitroxides on .NO homeostasis has not been studied yet. The present study investigates the effect of nitroxides on the detectable amount of .NO released by 3-morpholinosydnonimine (SIN-1) and cultured endothelial cells. Cultured bovine aortic and atrial endothelial cells stimulated with 10 microM A23187 released a stable flux of .NO, as detected by .NO chemiluminescence. Addition of 100 units/ml SOD or 10 microM of the nitroxides 4-hydroxy-2,2,6, 6-tetramethylpiperidine-N-oxyl (TEMPOL), 3-carboxy-proxyl, and 3-ethoxycarbonyl-proxyl, increased the chemiluminescence signal. The effect of these nitroxides on the amount of .NO released from cell monolayers was dose-dependent, with the highest efficacy between 30 and 100 microM. EPR spin trapping in SIN-1 solutions revealed the formation of .OH adducts from spontaneous dismutation of O-2 and concomitant reaction with H2O2. Both SOD and TEMPOL increased the signal intensity of the .OH adduct by accelerating the dismutation of O-2. The results of this study demonstrate that the SOD-mimetic activity of nitroxides increases the amount of bioavailable .NO in vitro.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
253324
3-Carboxy-PROXYL, 98%
C9H16NO3