EMAIL THIS PAGE TO A FRIEND
Journal of hepatology
Acute hepatitis caused by alverine associated with anti-lamin A and C autoantibodies.
PMID 9288616
Abstract
We report the case of a 67-year-old woman in whom onset and regression of acute hepatitis were closely related to the time of administration and withdrawal of the smooth muscle relaxant alverine. Antinuclear antibodies were positive, and their titer followed the course of hepatitis. They presented a smooth rim-like nuclear immunofluorescence staining pattern. Immunoblot assay showed that they were directed against lamin A and lamin C. This suggests that alverine should be added to the list of drugs known to produce acute hepatitis, and that drug-induced liver injury is a possible cause of antinuclear antibodies specific for lamin A and lamin C.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A0424
Alverine citrate salt
C20H27N · C6H8O7
Y0000687
Alverine citrate salt, European Pharmacopoeia (EP) Reference Standard
C20H27N · C6H8O7