EMAIL THIS PAGE TO A FRIEND
The Annals of pharmacotherapy
Zafirlukast for chronic asthma: convenient and generally safe, but is it effective?
PMID 9296252
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
Z4152
Zafirlukast, ≥98% (HPLC)
C31H33N3O6S