EMAIL THIS PAGE TO A FRIEND
Inflammation research : official journal of the European Histamine Research Society ... [et al.]
2,6-di-tert-butylphenols: a new class of potent and selective PGHS-2 inhibitor.
PMID 9297551
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D48400
2,6-Di-tert-butylphenol, 99%
C14H22O