EMAIL THIS PAGE TO A FRIEND
Journal of chromatography. A
Rapid pharmacokinetic screening of salbutamol in plasma samples by column-switching high-performance liquid chromatography-electrospray mass spectrometry.
PMID 9297839
Abstract
In order to obtain pharmacokinetic data from studies in humans, a sensitive and selective assay for the quantification of salbutamol in human plasma samples was required. This report describes an automated high-performance liquid chromatography-mass spectrometry assay with pre-column enrichment using internal standard calibration for the quantification of salbutamol and the validation of the assay. The lower limit of quantitation is 0.2 ng/ml with an accuracy and imprecision of less than 7%. The analysis time is 8 min per sample.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
S8260
Salbutamol
C13H21NO3