EMAIL THIS PAGE TO A FRIEND
Molecular and cellular biochemistry
The role of mitochondrial dysfunction and neuronal nitric oxide in animal models of neurodegenerative diseases.
PMID 9309687
Abstract
Excitotoxicity, mitochondrial dysfunction and free radical induced oxidative damage have been implicated in the pathogenesis of several different neurodegenerative diseases, such as amyotrophic lateral sclerosis, Parkinson's disease (PD), Alzheimer's disease (AD), and Huntington's disease. Much of the interest in the association of neurodegeneration with mitochondrial dysfunction and oxidative damage emerged from animal studies using mitochondrial toxins. Within mitochondria 1-methyl-4-phenylpyridinium (MPP+), the active metabolite of 1-methyl-4-phenyl-1,2,3,6-tetrahydropyridine (MPTP), acts to inhibit NADH-coenzyme Q reductase (complex I) of the electron transport chain. MPTP produces Parkinsonism in humans, primates, and mice. Similarly, lesions produced by the reversible inhibitor of succinate dehydrogenase (complex II), malonate, and the irreversible inhibitor, 3-nitropropionic acid (3-NP), closely resemble the histologic, neurochemical and clinical features of HD in both rats and non-human primates. The interruption of oxidative phosphorylation results in decreased levels of ATP. A consequence is partial neuronal depolarization and secondary activation of voltage-dependent NMDA receptors, which may result in excitotoxic neuronal cell death (secondary excitotoxicity). The increase in intracellular Ca2+ concentration leads to an activation of Ca2+ dependent enzymes, including the constitutive neuronal nitric oxide synthase (cnNOS) which produces NO.. NO. may react with the superoxide anion to from peroxynitrite. We show that systemic administration of 7-nitroindazole (7-NI), a relatively specific inhibitor of cnNOS in vivo. attenuates lesions produced by striatal malonate injections or systemic treatment with 3-NP or MPTP. Furthermore 7-NI attenuated increases in lactate production and hydroxyl radical and 3-nitrotyrosine generation in vivo, which may be a consequence of peroxynitrite formation. Our results suggest that neuronal nitric oxide synthase inhibitors may be useful in the treatment of neurologic diseases in which excitotoxic mechanisms play a role.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
N5636
3-Nitropropionic acid, ≥97%
C3H5NO4