EMAIL THIS PAGE TO A FRIEND
The Journal of antibiotics
Bestatin, an inhibitor of aminopeptidase B, produced by actinomycetes.
PMID 931798
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
B8385
Bestatin hydrochloride, ≥98% (HPLC)
C16H24N2O4 · HCl