EMAIL THIS PAGE TO A FRIEND
Contact dermatitis
Photoallergic contact dermatitis due to flufenamic acid and etofenamate.
PMID 9330834
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
E2499900 Etofenamate, European Pharmacopoeia (EP) Reference Standard
C18H18F3NO4