EMAIL THIS PAGE TO A FRIEND
European journal of pharmacology
Effect of selected triterpenoids on chronic dermal inflammation.
PMID 9346335
Abstract
The activity of four natural triterpenoids on a 12-O-tetradecanoylphorbol-13-acetate multiple-dose model of skin chronic inflammation was studied. Erythrodiol and ursolic acid were significantly effective. The most important features concerning structure-activity relationship and previous data on the effect of these triterpenoids on other inflammatory conditions are discussed.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
09258
Erythrodiol, analytical standard
C30H50O2