EMAIL THIS PAGE TO A FRIEND
Gan to kagaku ryoho. Cancer & chemotherapy
[Development of new drugs for urological cancers].
PMID 9350239
Abstract
Urological cancers have a variety of biological characteristics. Thus, anti-cancer agents in this field are often developed focusing on the biological characteristics. For example, LH-RH agonist and anti-androgens have been developed for androgen-sensitive prostate cancer, interferons and IL-2 for renal cell cancer, and BCG for superficial bladder cancer. In this manuscript, new agents which are thought to be promising in the urological field, bropirimine for bladder cancer and liarozole for prostate cancer, are briefly introduced.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML0218
Bropirimine, ≥98% (HPLC)
C10H8BrN3O