EMAIL THIS PAGE TO A FRIEND
Letters in applied microbiology
PCR amplification of crude microbial DNA extracted from soil.
PMID 9351282
Abstract
A rapid, inexpensive, large-scale DNA extraction method involving minimal purification has been developed that is applicable to various soil types. DNA was extracted from 100 g of soil using direct lysis with glass beads and sodium dodecyl sulphate (SDS) followed by polyethylene glycol precipitation, potassium acetate precipitation, phenol extraction and isopropanol precipitation. The crude extract could be used in PCR directed at high-copy number (bacterial small subunit rRNA) and single-copy (fungal beta-tubulin) genes.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P5708
Potassium acetate, BioXtra, ≥99.0%
C2H3KO2