EMAIL THIS PAGE TO A FRIEND
Methods in cell biology
A simple assay for local heating by optical tweezers.
PMID 9352510
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
85720
Butyl stearate, technical, 40-60% (GC)
C22H44O2