EMAIL THIS PAGE TO A FRIEND
Journal of bacteriology
Involvement of two alpha-ketoglutarate-dependent dioxygenases in enantioselective degradation of (R)- and (S)-mecoprop by Sphingomonas herbicidovorans MH.
PMID 9352915
Abstract
Cell extracts of Sphingomonas herbicidovorans MH grown on (R)-mecoprop contained an enzyme activity that selectively converted (R)-mecoprop to 4-chloro-2-methylphenol, whereas extracts of cells grown on (S)-mecoprop contained an enzyme activity selective for the S enantiomer. Both reactions were dependent on alpha-ketoglutarate and ferrous ions. Besides 4-chloro-2-methylphenol, pyruvate and succinate were detected as products of the reactions. Labeling experiments with (18)O2 revealed that both enzyme activities catalyzed a dioxygenation reaction. One of the oxygen atoms of pyruvate and one of the oxygen atoms of succinate were derived from molecular oxygen. Analysis of cell extracts obtained from cells grown on different substrates by sodium dodecyl sulfate-polyacrylamide gel electrophoresis showed that growth on (R)-mecoprop and (S)-mecoprop caused the appearance of prominent protein bands at 34 and 32 kDa, respectively. Both protein bands were present when cells grew on the racemic mixture. The results demonstrate that S. herbicidovorans initiated the degradation of each enantiomer of mecoprop by a specific alpha-ketoglutarate-dependent dioxygenase. By comparing conversion rates of various phenoxy herbicides, we confirmed that the two enzyme activities were distinct from that of TfdA, which catalyzes the first step in the degradation of 2,4-dichlorophenoxyacetic acid in Ralstonia eutropha JMP134.