EMAIL THIS PAGE TO A FRIEND
Cellular and molecular biology (Noisy-le-Grand, France)
Water soluble free radicals as biologically responsive agents in electron paramagnetic resonance imaging.
PMID 9359628
Abstract
Electron paramagnetic resonance imaging (EPRI) is currently being developed at frequencies between 200 MHz and 2 GHz. EPRI can map the in vivo distribution of paramagnetic species, such as water soluble free radicals; nitroxide free radicals are commonly used. EPR images reflect the complexity of metabolic actions on the exogenous delivered nitroxides. Their reduction rate in vivo is affected by parameters such as oxygen concentration, pH and biodistribution. This paper illustrates the main features of low frequency EPRI and reconstruction techniques. Examples of EPR imaging, such as two-dimensional (2D) spatial mapping of the distribution of a nitroxide free radical in phantoms and in whole rat, are given.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
253324
3-Carboxy-PROXYL, 98%
C9H16NO3