EMAIL THIS PAGE TO A FRIEND
Comparative biochemistry and physiology. Part C, Pharmacology, toxicology & endocrinology
Effect of 3-amino-1-propanol on the phosphatidylinositol (PI) and glycosyl phosphatidylinositol (GPI) systems of Tetrahymena.
PMID 9366039
Abstract
3-Amino-1-propanol (AP), a substance replacing ethanolamine in phosphatidylethanolamine (PE) significantly reduced 32P incorporation to phosphatidylinositol (PI) and glycosyl-phosphatidylinositol (GPI) in the unicellular organism Tetrahymena pyriformis. At 10 mM, AP completely inhibited the incorporation of 32P into PI. 3H-arachidonate incorporation into PI was also inhibited, while that into diacylglycerol (DAG) was high. The experiments indicate the presence and metabolism of inositol phospholipids and GPI in T. pyriformis.
Related Materials