EMAIL THIS PAGE TO A FRIEND
Biochimica et biophysica acta
Cyanate causes depletion of ascorbate in organisms.
PMID 9367185
Abstract
Ascorbate-dehydroascorbate redox cycle plays a key role in protecting organisms from an excess of oxidants. Recently, we found a novel reaction of dehydroascorbate with cyanate under the conditions of neutral pH and ordinary temperature. In this report, we demonstrated that through this irreversible reaction, cyanate causes the depletion of ascorbate in the matrix, where the ascorbate-dehydroascorbate redox cycle revolves. When the leaves of weed (Erigeron canadensis) were soaked in sodium cyanate solution generally used as a herbicide, the depletion of ascorbate as well as dehydroascorbate in them was observed, followed by the change in color from green to brown. These results suggest that a possible way of cyanate toxicity is to inflict oxidative stress on organisms.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
185086
Sodium cyanate, 96%
CNNaO