EMAIL THIS PAGE TO A FRIEND
Journal of virology
Early E-selectin, VCAM-1, ICAM-1, and late major histocompatibility complex antigen induction on human endothelial cells by flavivirus and comodulation of adhesion molecule expression by immune cytokines.
PMID 9371591
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
ECM645 Endothelial Cell Adhesion Assay Kit