EMAIL THIS PAGE TO A FRIEND
Blood
Inhibition of interferon regulatory factor-1 expression results in predominance of cell growth stimulatory effects of interferon-gamma due to phosphorylation of Stat1 and Stat3
PMID 9389691