EMAIL THIS PAGE TO A FRIEND
The Biochemical journal
Excess putrescine accumulation inhibits the formation of modified eukaryotic initiation factor 5A (eIF-5A) and induces apoptosis.
PMID 9396730
Abstract
DH23A cells, an alpha-difluoromethylornithine-resistant variant of the parental hepatoma tissue culture cells, express high levels of stable ornithine decarboxylase. Aberrantly high expression of ornithine decarboxylase results in a large accumulation of endogenous putrescine and increased apoptosis in DH23A cells when alpha-difluoromethylornithine is removed from the culture. Treatment of DH23A cells with exogenous putrescine in the presence of alpha-difluoromethylornithine mimics the effect of drug removal, suggesting that putrescine is a causative agent or trigger of apoptosis. Accumulation of excess intracellular putrescine inhibits the formation of hypusine in vivo, a reaction that proceeds by the transfer of the butylamine moiety of spermidine to a lysine residue in eukaryotic initiation factor 5A (eIF-5A). Treatment of DH23A cells with diaminoheptane, a competitive inhibitor of the post-translational modification of eIF-5A, causes both the suppression of eIF-5A modification in vivo and induction of apoptosis. These data support the hypothesis that rapid degradation of ornithine decarboxylase is a protective mechanism to avoid cell toxicity from putrescine accumulation. Further, these data suggest that suppression of modified eIF-5A formation is one mechanism by which cells may be induced to undergo apoptosis.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D17408
1,7-Diaminoheptane, 98%
C7H18N2