EMAIL THIS PAGE TO A FRIEND
Journal of pharmaceutical and biomedical analysis
Flow extraction spectrophotometric method for the determination of diclofenac sodium in pharmaceutical preparations.
PMID 9408840
Abstract
The spectrophotometric determination of trace amounts of diclofenac was carried out by liquid-liquid extraction using acridine yellow with a flow system. The determination of diclofenac sodium in the range of 3-80 micrograms ml-1 was possible with a sampling frequency of 40 samples h-1. The method was satisfactorily applied to the determination of diclofenac in pharmaceutical preparations.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
199508
Acridine Yellow G, Dye content 90 %
C15H15N3 · HCl