EMAIL THIS PAGE TO A FRIEND
American journal of respiratory and critical care medicine
Inhalation of the nitric oxide synthase cofactor tetrahydrobiopterin in healthy volunteers.
PMID 9412588
Abstract
Pulmonary endothelial dysfunction is the hallmark of acute lung injury. Impaired pulmonary endothelial nitric oxide (NO) production in this event has been described. Tetrahydrobiopterin (BH4) is an essential cofactor for NO synthase and modulator of its activity. At high local concentrations, BH4 provokes local vasodilation in vivo in healthy individuals. At lower concentrations, BH4 selectively and locally restores disturbed NO-dependent vasodilation in patients with endothelial dysfunction. In this preliminary study, we therefore investigated the feasibility of BH4 inhalation in five healthy human volunteers. Inhalation of buffered, aqueous BH4-dihydrochloride solution was well tolerated; despite the buffer, BH4 stability was completely preserved. Resorption of inhaled BH4 was demonstrated by significantly increased BH4 levels in plasma and urine. Inhaled BH4 did not alter pulmonary function and had no effect on systemic hemodynamic values. Our data demonstrate that inhalation is a novel method for local BH4 administration, offering a basic therapeutic tool for investigation of restoration of impaired NO-dependent vasodilation due to pulmonary endothelial dysfunction.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
I7388
Isoxanthopterin, ≥97.5%, powder
C6H5N5O2