EMAIL THIS PAGE TO A FRIEND
Agents and actions
A method for the measurement of ultra-low amounts of prostaglandins in inflammatory exudates using gas chromatography and mass spectrometry.
PMID 941791
Abstract
A GC-MS assay for PGE2 and PGF2alpha has been described. The method is highly specific and sensitive down to the sub-picogram level. The assay has been used to measure the time dependence of PG concentration in carrageenan-induced inflammatory exudates and the results compared with a bioassay. Estimates of the extraction and purification yield of PGs from exudates have also been obtained.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
33031-U BSA+TMCS+TMSI, 3:2:3, pkg of 1 × 25 mL