EMAIL THIS PAGE TO A FRIEND
Analytical biochemistry
A simple and highly sensitive mass fragmentographic procedure for gamma-aminobutyric acid determinations.
PMID 942039
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
52619
Hexamethyldisilazane, for GC derivatization, ≥99.0% (GC)
C6H19NSi2