EMAIL THIS PAGE TO A FRIEND
Nucleic acids research
Electrospray ionization mass spectrometric characterization of photocrosslinked DNA-EcoRI DNA methyltransferase complexes.
PMID 9421528
Abstract
We describe a novel strategy combining photocrosslinking and HPLC-based electrospray ionization mass spectrometry to identify UV crosslinked DNA-protein complexes. Eco RI DNA methyltransferase modifies the second adenine within the recognition sequence GAATTC. Substitution of 5-iodouracil for the thymine adjacent to the target base (GAATTC) does not detectably alter the DNA-protein complex. Irradiation of the 5-iodouracil-substituted DNA-protein complex at various wavelengths was optimized, with a crosslinking yield >60% at 313 nm after 1 min. No protein degradation was observed under these conditions. The crosslinked DNA-protein complex was further analyzed by electrospray ionization mass spectrometry. The total mass is consistent with irradiation-dependent covalent bond formation between one strand of DNA and the protein. These preliminary results support the possibility of identifying picomole quantities of crosslinked peptides by similar strategies.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
857858
5-Iodouracil, 98%
C4H3IN2O2