EMAIL THIS PAGE TO A FRIEND
Drug metabolism reviews
Evaluation of gastrointestinal absorption and metabolism.
PMID 9421681
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
1000420 Abacavir Related Compound A, United States Pharmacopeia (USP) Reference Standard
C11H14N6O