EMAIL THIS PAGE TO A FRIEND
Indian journal of biochemistry & biophysics
Rubiadin, a new antioxidant from Rubia cordifolia.
PMID 9425750
Abstract
Rubiadin, a dihydroxy anthraquinone, isolated from alcoholic extract of Rubia cordifolia, possesses potent antioxidant property. It prevents lipid peroxidation induced by FeSO4 and t-butylhydroperoxide (t-BHP) in a dose dependent manner. The per cent inhibition was more in the case of Fe2+ induced lipid peroxidation. The antioxidant property of the preparation has been found to be better than that of EDTA, Tris, mannitol, Vitamin E and p-benzoquinone.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
74553
Rubiadin, analytical standard
C15H10O4