EMAIL THIS PAGE TO A FRIEND
Journal of chromatographic science
Mefanamic acid blood and urine levels in the horse determined by derivative gas-liquid chromatography-electron capture.
PMID 943417
Abstract
Mefenamic acid is extracted from biological fluids and is acylated with pentafluoropropionic anhydride to form a derivative possessing high electron affinity. The derivative is analyzed by gas-liquid chromatography with an electron capture detector. The method is particularly valuable for determining drug levels in blood where small sample and/or drug concentrations are available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
details
Pentafluoropropionic anhydride
C6F10O3