EMAIL THIS PAGE TO A FRIEND
The New England journal of medicine
Respiratory function of hemoglobin.
PMID 9435331
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
H4890 Anti-Human Hemoglobin antibody produced in rabbit, whole antiserum