EMAIL THIS PAGE TO A FRIEND
Pediatric nephrology (Berlin, Germany)
Renal stones in nephropathic cystinosis treated with phosphocysteamine.
PMID 9438669
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C8397
Cysteamine S-phosphate sodium salt, analytical standard
C2H7NNaO3PS